Learn More
Medchemexpress LLC Arc 239 dihydrochloride | 55974-42-0 | MFCD02262215; MFCD16875414 | 99.2% | 480.43 | C24H31Cl2N3O3 | 5 MG
SDP
Catalog No. 5000264857
Encompass_Preferred
ARC 239 dihydrochloride is a small-molecule research compound that acts as a selective α2B/α2C adrenoceptor antagonist. It is supplied as a white to off-white solid intended for in vitro pharmacology and receptor-binding studies; reported binding data indicate greater affinity at α2B compared with α2A and α2C subtypes.
  • Selective antagonist of α2B and α2C adrenoceptors.
  • Reported pKd/pKi values indicate higher affinity for α2B subtype.
  • High purity suitable for research applications.
  • White to off-white solid form for convenient handling.
  • Stable under recommended refrigerated storage.
  • Useful for subtype-selective adrenergic receptor studies.
Catalog No. 50-002-64857 Supplier Medchemexpress LLC Supplier No. HY12709A5MG
ARC 239 dihydrochloride is a small-molecule research compound that acts as a selective α2B/α2C adrenoceptor antagonist. It is supplied as a white to off-white solid intended for in vitro pharmacology and receptor-binding studies; reported binding data indicate greater affinity at α2B compared with α2A and α2C subtypes.
  • Selective antagonist of α2B and α2C adrenoceptors.
  • Reported pKd/pKi values indicate higher affinity for α2B subtype.
  • High purity suitable for research applications.
  • White to off-white solid form for convenient handling.
  • Stable under recommended refrigerated storage.
  • Useful for subtype-selective adrenergic receptor studies.