Learn More
National Safety Apparel™ Arc Flash Hood 7.5 oz. Navy Tecgen CC With Quilted Liner
Shop All National Safety Apparel Products
Specifications
Specifications
| Size | Medium |
| Material | Rayon, Modacrylic, O-Pan, Para-Aramid |
| Color | Navy |
| Seam Type | Stitched |
| Arc Flash Rating | 55 cal/cm² |
| Certifications/Compliance | CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269 |