Learn More
Honeywell Salisbury™ Arc Flash Kit
Catalog No. 19139209 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
8 cal/cm²
Description:
46 to 48 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back packs and safety Glasses
50 to 52 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses
Includes: 42 to 44 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating Description Size
19-139-209 8 cal/cm² 50 to 52 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses 2X-Large
19-139-207 8 cal/cm² Includes: 42 to 44 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses Large
19-139-208 8 cal/cm² 46 to 48 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back packs and safety Glasses X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-139-209 Supplier Honeywell Salisbury™ Supplier No. SKCA82XLBP
This item is not returnable. View return policy
Specifications
Arc Flash Rating 8 cal/cm²
Size 2X-Large
Description 50 to 52 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses
Certifications/Compliance NFPA 70E™ 2009, CE 0120
Color Navy
Material Cotton