Learn More
MSC PIP Fire-Resistant Vest, Solid Westex® DH, Hi-Vis Yellow, 2XL, 1 ea
Catalog No. 5000688666
Encompass
This solid arc-rated and flame-resistant vest is designed for construction and municipal work environments requiring high-visibility protection combined with arc flash safety. It is engineered to be worn exclusively over other arc-rated clothing to provide additional visibility and protection. The garment utilizes a durable, long-lasting fabric construction and complies with rigorous safety standards for use in hazardous areas.
  • Arc rating of 8.9 cal/cm2 (PPE 2)
  • Meets NFPA 70E, ASTM F1506, and ANSI/ISEA 107-2020 Type R Class 3 requirements
  • Features 2 in. retroreflective silver flame-resistant tape for enhanced visibility
  • Constructed from durable Westex® DH fabric
  • Equipped with a flame-resistant hook and loop closure for secure donning and doffing
Catalog No. 50-006-88666 Supplier MSC Supplier No. 95448692
Add to Cart
Add to Cart
This solid arc-rated and flame-resistant vest is designed for construction and municipal work environments requiring high-visibility protection combined with arc flash safety. It is engineered to be worn exclusively over other arc-rated clothing to provide additional visibility and protection. The garment utilizes a durable, long-lasting fabric construction and complies with rigorous safety standards for use in hazardous areas.
  • Arc rating of 8.9 cal/cm2 (PPE 2)
  • Meets NFPA 70E, ASTM F1506, and ANSI/ISEA 107-2020 Type R Class 3 requirements
  • Features 2 in. retroreflective silver flame-resistant tape for enhanced visibility
  • Constructed from durable Westex® DH fabric
  • Equipped with a flame-resistant hook and loop closure for secure donning and doffing