Learn More
National Safety Apparel™ Arc H2O FR Hi-Vis Orange Bib Overall - Class E
SDP
Catalog No. 170012204 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2204 5X-Large
17-001-2197 Small
17-001-2198 Medium
17-001-2199 Large
17-001-2200 2X-Large
17-001-2201 3X-Large
17-001-2202 4X-Large
17-001-2203 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2204 Supplier National Safety Apparel™ Supplier No. R40RQ145X
Arc H2O FR HI-Vis rain bib overall class E
  • Arc Rating: 19 cal/cm² (Fluorescent Yellow), 24 cal/cm² (Fluorescent Orange)
  • CAT 2
  • Fabric/Material: PU Coated FR Cotton
  • Waterproof welded seams with reinforced stress points to provide durable construction
  • FR fabric will not melt, drip or ignite
  • FR polyurethane is durable and flexible that will not become brittle as it ages
  • Convenient snap fly pant front
  • Adjustable shoulder straps
  • 2 in. silver reflective trim
  • Adjustable elastic suspender system for fit and comfort
  • Double storm flap that keeps weather out
  • FR hook and loop tabs to adjust leg cuff
  • Made in the USA
Specifications
Closure Type Buckle
Color Fluorescent Orange
Material Polyurethane-coated Cotton
Seam Type Welded
Size 5X-Large
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange)
Certifications/Compliance CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Disposable Reusable