Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Arc H2O™ FR Hi-Vis Orange Trench Coat, Type R Class 3
SDP

Catalog No. 170012156 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2156 5X-Large
17-001-2149 X-Large
17-001-2150 Small
17-001-2151 Medium
17-001-2152 Large
17-001-2153 2X-Large
17-001-2154 3X-Large
17-001-2155 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2156 Supplier National Safety Apparel™ Supplier No. R31RQ065X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arc H2O FR HI-Vis rain trench coat type R class 3

  • 48 in. trench coat provides superior garment coverage
  • Waterproof welded seams with reinforced stress points to provide durability
  • FR fabric will not melt, drip or ignite
  • FR polyurethane is durable and flexible that will not become brittle as it ages
  • Contoured collar with tuck away attached hood
  • Front storm flap with reflective trim improves comfort and visibility
  • FR hook and loop tab at wrist cuff for improved fit
  • Back ventilation system with FR mesh for increased airflow
  • Covered D-Ring opening access
  • Two reinforced front pockets with closure for convenient secure storage
  • 2 in. silver reflective trim
  • Adjustable cuffs with hook and loop tabs
  • Reinforced stress points provides durable construction
  • Covered zipper and snap closures
  • Made in the USA

Specifications

Size 5X-Large
Gender Unisex
Material Polyurethane-coated Cotton
Color Fluorescent Orange
Closure Type Zipper
Certifications/Compliance Arc Rating: 9.8 cal/cm² , CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Seam Type Welded
Disposable Reusable
Product Type Hi-vis Rain Jacket
Style Jacket
For Use With (Application) Utilities, Electrical Contractors
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.