Learn More
Waters Corp Performance Maintenance Kit, ACQUITY Arc Quaternary Solvent Manager, Annual
Catalog No. 502392974 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the ACQUITY Arc Quaternary Solvent Manager is designed to maintain the optimal performance of the instrument on an annual basis. This kit includes essential Waters Quality Parts™ to ensure reliable operation and longevity of the system.
  • Specifically designed for ACQUITY Arc Quaternary Solvent Manager instruments.
  • Provides all necessary parts for annual maintenance.
  • Ensures peak performance and reliable operation.
  • Includes mixers, plungers, check valves, seals, and filters.
  • Supports UPLC system types.
Catalog No. 50-239-2974 Supplier Waters Corp Supplier No. 201000317
The Waters™ Performance Maintenance Kit for the ACQUITY Arc Quaternary Solvent Manager is designed to maintain the optimal performance of the instrument on an annual basis. This kit includes essential Waters Quality Parts™ to ensure reliable operation and longevity of the system.
  • Specifically designed for ACQUITY Arc Quaternary Solvent Manager instruments.
  • Provides all necessary parts for annual maintenance.
  • Ensures peak performance and reliable operation.
  • Includes mixers, plungers, check valves, seals, and filters.
  • Supports UPLC system types.