Learn More
Grainger Arc Screen 72 W x 2 D x 48 H Unpainted
Catalog No. 502767442
Encompass_Preferred
The PRO-LINE Arc Screen is a portable welding screen that protects from welding sparks and UV/IR light. It features flame-retardant vinyl with bound edges to prevent fraying. The screen attaches to a lightweight yet sturdy aluminum frame, supporting a 52" x 34" viewing area. The frame assembles easily with snap buttons and folds compactly for storage and transport. Ideal for MIG, TIG, stick, and oxy-fuel welding, grinding, and plasma cutting, it protects the welder's eyes and face while containing sparks and slag. The PRO-LINE Arc Screen is an affordable and practical welding safety solution.
Catalog No. 50-276-7442 Supplier Grainger Supplier No. 16Y921
The PRO-LINE Arc Screen is a portable welding screen that protects from welding sparks and UV/IR light. It features flame-retardant vinyl with bound edges to prevent fraying. The screen attaches to a lightweight yet sturdy aluminum frame, supporting a 52" x 34" viewing area. The frame assembles easily with snap buttons and folds compactly for storage and transport. Ideal for MIG, TIG, stick, and oxy-fuel welding, grinding, and plasma cutting, it protects the welder's eyes and face while containing sparks and slag. The PRO-LINE Arc Screen is an affordable and practical welding safety solution.