Learn More
Fauske & Associates, a Division of Stone & Webster, LLC ARC TEST CELL FH-4 SIDE CLIP
Catalog No. NC3451907
Encompass
ARC Test Cellsall metal, long neck, Hastelloy C, 1" I.D., .025 wall with 1/4" O.D. x .035 wall x 1" stems, side clipthese cells can be used with the Accelerating Rate Calorimeter (ARC) and the ARSST
Catalog No. NC3451907 Supplier Fauske & Associates, a Division of Stone & Webster, LLC Supplier No. ARCFH4SC
ARC Test Cellsall metal, long neck, Hastelloy C, 1" I.D., .025 wall with 1/4" O.D. x .035 wall x 1" stems, side clipthese cells can be used with the Accelerating Rate Calorimeter (ARC) and the ARSST