Learn More
Fauske & Associates, a Division of Stone & Webster, LLC ARC TEST FH-1A SIDE CLIP
Catalog No. NC3451903
Encompass
ARC Test Cellsall metal, long neck, Hastelloy C - 1" I.D., .035 wall with 1/8" O.D. x .020 wall x 1" stems, side clip, volume: 9 mLthese cells can be used with the Accelerating Rate Calorimeter (ARC) and the ARSST
Catalog No. NC3451903 Supplier Fauske & Associates, a Division of Stone & Webster, LLC Supplier No. ARCFH1ASC
ARC Test Cellsall metal, long neck, Hastelloy C - 1" I.D., .035 wall with 1/8" O.D. x .020 wall x 1" stems, side clip, volume: 9 mLthese cells can be used with the Accelerating Rate Calorimeter (ARC) and the ARSST