Learn More
Apexbio Technology LLC ARCA, 100ul (100 mM). Cas: MFCD: NA. Anti Reverse Cap Analog (ARCA) used to forming Cap 0 structure,3´-O-Me-m7G(5')ppp(5')G
SDP
Catalog No. 501905482
Encompass_Preferred
A critical step in mRNA processing is the addition of a 5 cap structure, a 5'-5' triphosphate linkage between the 5' initiating terminal of the RNA and a guanosine nucleotide. The cap is then methylated enzymatically at the N-7 position of the guanosine to form a mature mCAP. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5482 Supplier Apexbio Technology LLC Supplier No. B8175.1
A critical step in mRNA processing is the addition of a 5 cap structure, a 5'-5' triphosphate linkage between the 5' initiating terminal of the RNA and a guanosine nucleotide. The cap is then methylated enzymatically at the N-7 position of the guanosine to form a mature mCAP. Other sizes are available. Please inqury us for quote.