Learn More
Apexbio Technology LLC ARCA Cy3 EGFP mRNA (5-moUTP), 100ug (1mg/mL)
SDP
Catalog No. 501998307
Encompass_Preferred
ARCA Cy3 EGFP mRNA (5-moUTP) can be used to analyze mRNA delivery and translation efficiency. The EGFP mRNA will express an enhanced version of green fluorescent protein, which was originally isolated from the jellyfish, Aequorea victoria. EGFP is a direct detection reporter gene commonly used in mammalian cell cultures to produce bright green fluorescence with an emission peak at 509 nm. EGFP mRNA labeled with cyanine 3 can be directly visualized. ARCA Cy3 EGFP mRNA (5-moUTP) is an ideal molecule for determining mRNA delivery and localization and is independent of translation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8307 Supplier Apexbio Technology LLC Supplier No. R1008.1
ARCA Cy3 EGFP mRNA (5-moUTP) can be used to analyze mRNA delivery and translation efficiency. The EGFP mRNA will express an enhanced version of green fluorescent protein, which was originally isolated from the jellyfish, Aequorea victoria. EGFP is a direct detection reporter gene commonly used in mammalian cell cultures to produce bright green fluorescence with an emission peak at 509 nm. EGFP mRNA labeled with cyanine 3 can be directly visualized. ARCA Cy3 EGFP mRNA (5-moUTP) is an ideal molecule for determining mRNA delivery and localization and is independent of translation. Other sizes are also available. Please inqury us for quote.