Learn More
Apexbio Technology LLC ARCA EGFP mRNA, 100ug (1mg/mL). Cas: MFCD: NA. Direct-detection reporter mRNA, used as control to study transfection and expression in mammalian cells.
SDP
Catalog No. 501905651
Encompass_Preferred
EGFP (enhanced green fluorescent protein) mRNA can express an enhanced green fluorescence once has been transfected into cells, which makes it always used as a direct detection reporter mRNA. You can observe the fluorescence at 509 nm. Our product had already been modified with ARCA with a high efficient co-transcriptional capping method, which means most of mRNA has a Cap 0 structure. It is a more mature mRNA than mRNA with no capping and maintain a more stable state which lead to higher expressing efficiency in transfected mammalian cells. ARCA obtain the ability only inserting in the proper orientation, results in forming mRNAs that can be translated twice efficiently as those initiated with mCAP. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5651 Supplier Apexbio Technology LLC Supplier No. R10010.1
EGFP (enhanced green fluorescent protein) mRNA can express an enhanced green fluorescence once has been transfected into cells, which makes it always used as a direct detection reporter mRNA. You can observe the fluorescence at 509 nm. Our product had already been modified with ARCA with a high efficient co-transcriptional capping method, which means most of mRNA has a Cap 0 structure. It is a more mature mRNA than mRNA with no capping and maintain a more stable state which lead to higher expressing efficiency in transfected mammalian cells. ARCA obtain the ability only inserting in the proper orientation, results in forming mRNAs that can be translated twice efficiently as those initiated with mCAP. Other sizes are available. Please inqury us for quote.