Learn More
Apexbio Technology LLC ARCA EGFP mRNA (5-moUTP), 100ug (1mg/mL)
SDP
Catalog No. 501998304
Encompass_Preferred
EGFP (enhanced green fluorescent protein) mRNA can express an enhanced green fluorescence when transfected into cells, it is used as a direct-detection reporter mRNA with fluorescence emission at 509 nm. Our ARCA EGFP mRNA (5-moUTP) is provided at a concentration of 1mg/ml, it is ARCA capped, polyA-tailed and incorporated with modified nucleotides.Anti-Reverse Cap Analog (ARCA) is a cap analog used during in vitro transcription for the generation of capped transcripts. ARCA obtains the ability only inserting in the proper orientation, results in forming mRNAs that can be translated twice efficiently as those initiated with mCAP. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8304 Supplier Apexbio Technology LLC Supplier No. R1007.1
EGFP (enhanced green fluorescent protein) mRNA can express an enhanced green fluorescence when transfected into cells, it is used as a direct-detection reporter mRNA with fluorescence emission at 509 nm. Our ARCA EGFP mRNA (5-moUTP) is provided at a concentration of 1mg/ml, it is ARCA capped, polyA-tailed and incorporated with modified nucleotides.Anti-Reverse Cap Analog (ARCA) is a cap analog used during in vitro transcription for the generation of capped transcripts. ARCA obtains the ability only inserting in the proper orientation, results in forming mRNAs that can be translated twice efficiently as those initiated with mCAP. Other sizes are also available. Please inqury us for quote.