Learn More
Medchemexpress LLC Arcaine sulfate | 14923-17-2 | MFCD00050545 | 98.0% | 270.31 | C6H18N6O4S | 5 MG
SDP
Catalog No. 5000211551
Encompass_Preferred
Arcaine sulfate is the sulfate salt of arcaine, a glutamate NMDA receptor antagonist used in neuroscience research. Supplied as a white to off-white solid with reported purity of 98.0%, it is intended for research use only and provides a tool to investigate NMDA receptor-mediated signaling in vitro and ex vivo. Store according to supplier recommendations to maintain stability.
  • Potent NMDA receptor antagonist for receptor pharmacology studies.
  • High purity (98.0%) for reliable experimental results.
  • White to off-white solid suitable for standard laboratory handling.
  • Compatible with in vitro and ex vivo neuroscience assays.
  • Recommended storage conditions provided to preserve compound integrity.
Catalog No. 50-002-11551 Supplier Medchemexpress LLC Supplier No. HY1009205MG
Arcaine sulfate is the sulfate salt of arcaine, a glutamate NMDA receptor antagonist used in neuroscience research. Supplied as a white to off-white solid with reported purity of 98.0%, it is intended for research use only and provides a tool to investigate NMDA receptor-mediated signaling in vitro and ex vivo. Store according to supplier recommendations to maintain stability.
  • Potent NMDA receptor antagonist for receptor pharmacology studies.
  • High purity (98.0%) for reliable experimental results.
  • White to off-white solid suitable for standard laboratory handling.
  • Compatible with in vitro and ex vivo neuroscience assays.
  • Recommended storage conditions provided to preserve compound integrity.