Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arcaine sulfate | 14923-17-2 | MFCD00050545 | 98.0% | 270.31 | C6H18N6O4S | 50 MG
SDP

Catalog No. 5000212625
Encompass_Preferred

Arcaine sulfate is the sulfate salt of Arcaine, a small-molecule glutamate N-methyl-D-aspartate (NMDA) receptor inhibitor supplied for research use in neuropharmacology. It is intended for in vitro and in vivo studies investigating NMDA receptor-mediated signaling and pharmacological modulation.

  • Glutamate NMDA receptor inhibitor.
  • CAS number 14923-17-2.
  • Molecular formula C6H18N6O4S.
  • Molecular weight 270.31.
  • Purity 98.0%.
  • Available in research sizes including 50 mg.

Catalog No. 50-002-12625 Supplier Medchemexpress LLC Supplier No. HY10092050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arcaine sulfate is the sulfate salt of Arcaine, a small-molecule glutamate N-methyl-D-aspartate (NMDA) receptor inhibitor supplied for research use in neuropharmacology. It is intended for in vitro and in vivo studies investigating NMDA receptor-mediated signaling and pharmacological modulation.

  • Glutamate NMDA receptor inhibitor.
  • CAS number 14923-17-2.
  • Molecular formula C6H18N6O4S.
  • Molecular weight 270.31.
  • Purity 98.0%.
  • Available in research sizes including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.