Learn More
Medchemexpress LLC Arcaine sulfate | 14923-17-2 | MFCD00050545 | 98.0% | 270.31 g/mol | C6H18N6O4S | 10 MG
SDP
Catalog No. 5000213359
Encompass_Preferred
Arcaine sulfate is the sulfate salt of arcaine, a small-molecule NMDA (N-methyl-D-aspartate) receptor antagonist used in neuroscience and pharmacology research. It is provided as a crystalline powder for laboratory use and is intended for in vitro pharmacology, receptor-binding assays, and studies of glutamatergic signaling mechanisms.
  • Glutamate NMDA receptor inhibitor for receptor and signaling studies.
  • High purity (98.0%) suitable for analytical and pharmacological assays.
  • Supplied as a sulfate salt to improve stability and handling.
  • Available in small pack sizes appropriate for lab-scale experiments.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality control and safe use.
Catalog No. 50-002-13359 Supplier Medchemexpress LLC Supplier No. HY10092010MG
Arcaine sulfate is the sulfate salt of arcaine, a small-molecule NMDA (N-methyl-D-aspartate) receptor antagonist used in neuroscience and pharmacology research. It is provided as a crystalline powder for laboratory use and is intended for in vitro pharmacology, receptor-binding assays, and studies of glutamatergic signaling mechanisms.
  • Glutamate NMDA receptor inhibitor for receptor and signaling studies.
  • High purity (98.0%) suitable for analytical and pharmacological assays.
  • Supplied as a sulfate salt to improve stability and handling.
  • Available in small pack sizes appropriate for lab-scale experiments.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality control and safe use.