Learn More
Medchemexpress LLC Arcaine sulfate | 14923-17-2 | MFCD00050545 | 98.0% | 270.31 g/mol | C6H18N6O4S | 100 MG
SDP
Catalog No. 5000212389
Encompass_Preferred
Arcaine sulfate is the sulfate salt of arcaine, a glutamate N-methyl-D-aspartate (NMDA) receptor inhibitor supplied as a research-grade reagent for neuroscience and pharmacology studies. It is provided as a crystalline sulfate salt with reported purity 98.0% and molecular weight 270.31 g/mol; for research use only.
  • Acts as an NMDA receptor antagonist for glutamatergic signaling studies.
  • High purity (98.0%) suitable for analytical and pharmacological assays.
  • Water-soluble sulfate salt with molecular weight 270.31 g/mol.
  • Available in multiple pack sizes for flexible experimental needs.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-12389 Supplier Medchemexpress LLC Supplier No. HY100920100MG
Arcaine sulfate is the sulfate salt of arcaine, a glutamate N-methyl-D-aspartate (NMDA) receptor inhibitor supplied as a research-grade reagent for neuroscience and pharmacology studies. It is provided as a crystalline sulfate salt with reported purity 98.0% and molecular weight 270.31 g/mol; for research use only.
  • Acts as an NMDA receptor antagonist for glutamatergic signaling studies.
  • High purity (98.0%) suitable for analytical and pharmacological assays.
  • Water-soluble sulfate salt with molecular weight 270.31 g/mol.
  • Available in multiple pack sizes for flexible experimental needs.
  • For research use only; not for human or veterinary use.