Learn More
Medchemexpress LLC Arcaine sulfate | 14923-17-2 | MFCD00050545 | 98.0% | 270.31 g/mol | C6H18N6O4S | 25 MG
SDP
Catalog No. 5000212267
Encompass_Preferred
Arcaine sulfate is the sulfate salt of a research-grade NMDA receptor antagonist used to modulate glutamatergic signaling in neuroscience and pharmacology studies. It is supplied as a high-purity solid suitable for in vitro and preclinical applications, with small pack sizes that support dose-ranging and reproducible experiments.
  • High purity (98.0%) suitable for research applications.
  • Acts as a glutamate NMDA receptor inhibitor for studies of excitatory neurotransmission.
  • Available in multiple small pack sizes for dose-ranging experiments.
  • Sulfate salt form improves stability and handling.
  • Molecular identity C6H18N6O4S enables reproducible results.
Catalog No. 50-002-12267 Supplier Medchemexpress LLC Supplier No. HY10092025MG
Arcaine sulfate is the sulfate salt of a research-grade NMDA receptor antagonist used to modulate glutamatergic signaling in neuroscience and pharmacology studies. It is supplied as a high-purity solid suitable for in vitro and preclinical applications, with small pack sizes that support dose-ranging and reproducible experiments.
  • High purity (98.0%) suitable for research applications.
  • Acts as a glutamate NMDA receptor inhibitor for studies of excitatory neurotransmission.
  • Available in multiple small pack sizes for dose-ranging experiments.
  • Sulfate salt form improves stability and handling.
  • Molecular identity C6H18N6O4S enables reproducible results.