Learn More
Medchemexpress LLC ARCC-4 | 1973403-00-7 | 99.9% | 1024.18 g·mol⁻1 | C53H56F3N7O7S2 | 5 MG
SDP
Catalog No. 5000263099
Encompass_Preferred
ARCC-4 is a PROTAC-based androgen receptor degrader intended for biochemical and cellular research. The molecule is enzalutamide-derived and recruits the von Hippel-Lindau E3 ligase to induce androgen receptor degradation at low nanomolar potency (reported DC50 ≈ 5 nM). Supplied as a dry powder with storage instructions for long-term stability.
  • Potent androgen receptor degrader with low-nanomolar activity.
  • Enzalutamide-derived VHL-recruiting PROTAC mechanism.
  • Active against clinically relevant androgen receptor mutants.
  • Provided as a dry powder suitable for biochemical and cellular assays.
  • Long-term storage stability: powder -20°C for 3 years, 4°C for 2 years.
  • High chemical purity for reproducible experimental results.
  • Available in small mass packages for research use.
Catalog No. 50-002-63099 Supplier Medchemexpress LLC Supplier No. HY1304925MG
ARCC-4 is a PROTAC-based androgen receptor degrader intended for biochemical and cellular research. The molecule is enzalutamide-derived and recruits the von Hippel-Lindau E3 ligase to induce androgen receptor degradation at low nanomolar potency (reported DC50 ≈ 5 nM). Supplied as a dry powder with storage instructions for long-term stability.
  • Potent androgen receptor degrader with low-nanomolar activity.
  • Enzalutamide-derived VHL-recruiting PROTAC mechanism.
  • Active against clinically relevant androgen receptor mutants.
  • Provided as a dry powder suitable for biochemical and cellular assays.
  • Long-term storage stability: powder -20°C for 3 years, 4°C for 2 years.
  • High chemical purity for reproducible experimental results.
  • Available in small mass packages for research use.