Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 Cal Long Coat/Legging Kit with Balaclava/Voltage Gloves (08)
SDP

Catalog No. 170011169 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1169 Small
17-001-1174 Medium
17-001-1179 Large
17-001-1184 X-Large
17-001-1189 2X-Large
17-001-1194 3X-Large
17-001-1199 4X-Large
17-001-1204 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1169 Supplier National Safety Apparel™ Supplier No. KIT2LC11SM08
Add to Cart
Add to Cart

Ultimate HRC 2 kit without balaclava and gloves

  • Coveralls
  • Hard Hat with ArcGuard™ Faceshield & Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Gear bag

Specifications

Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Gender Unisex
Material Cotton, Nylon
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Arc Flash Kit
Arc Flash Rating 12 cal/cm²
Seam Type Stitched
Size Small
Closure Type FR Hook and Loop
Color Navy
Suspender None
Ankle Style Open
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.