Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 Cal Coverall Kit with Voltage Gloves (11)
SDP

Catalog No. 170011001 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1001 Medium
17-001-1006 Large
17-001-1011 X-Large
17-001-1016 2X-Large
17-001-1021 3X-Large
17-001-1026 4X-Large
17-001-1031 5X-Large
17-001-996 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1001 Supplier National Safety Apparel™ Supplier No. KIT2CV20MD11
Add to Cart
Add to Cart

20 cal/cm² Level 2 Arc Flash Kit with an FR Coverall in UltraSoft

  • 20 cal FR Coverall made from UltraSoft™
  • 11 in. Class 0 Rubber Voltage Insulating Gloves
  • 10 in. Leather Glove Protectors
  • Hard Hat with Arc Flash Faceshield and Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 20 cal/cm²
Size Medium
Color Navy
Ankle Style Open
Closure Type Zipper
Material Westex™ UltraSoft™
Seam Type Stitched
Suspender None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.