Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 cal Coat/Bib Kit with Lift Front Hood/Voltage Gloves (08)
SDP

Catalog No. p-7220160 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1334 Small
17-001-1339 Medium
17-001-1344 Large
17-001-1349 X-Large
17-001-1354 2X-Large
17-001-1359 3X-Large
17-001-1364 4X-Large
17-001-1369 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1334 Supplier National Safety Apparel™ Supplier No. KIT2SC20LFSM08
Only null left
Add to Cart
Add to Cart

The premium choice for superior comfort, improved safety and above average durability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots

Specifications

Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 20 cal/cm²
Color Navy
Closure Type FR Hook and Loop
Ankle Style Open
Size Small
Material Westex™ UltraSoft™
Seam Type Stitched
Suspender Adjustable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.