Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ 25 kA ArcGuard™ Suppression Blanket, 4 ft x 5 ft
SDP

Catalog No. 17001752 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
48 x 60 in.
48 x 96 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-001-752 48 x 96 in.
17-001-751 48 x 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-001-752 Supplier National Safety Apparel™ Supplier No. K25LB4F8F
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal solution for jobs in confined and enclosed spaces where arc flash is a hazard. It is available on its own, or in a convenient kit package which includes the blanket, straps, and carrying bags

  • 12 attachment points
  • Multi-layer material with reinforced holes for straps
  • Provides optimal barrier in enclosed and confined spaces
  • Breakopen Threshold Performance (BTP) = 362 kA cycles
  • Maximum Arc current Imax = 25 kA
  • Made in the USA

Specifications

Product Type Fire Blanket
Color Navy
Material Multi-layer Material with Reinforced Holes
Certifications/Compliance NFPA 70 E, ASTM F2676, CSA Z462, OSHA 1910.269, Maximum Arc current Imax = 25 kA
Dimensions (L x W) 48 x 96 in.
Disposable Limited-use
For Use With (Application) Electrical Contractors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.