Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 40 cal Contractor Arc Flash Kit with Short Coat and Bib Overall (08)
SDP

Catalog No. 170011697 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1697 5X-Large
17-001-1662 Small
17-001-1667 Medium
17-001-1672 Large
17-001-1677 X-Large
17-001-1682 2X-Large
17-001-1687 3X-Large
17-001-1692 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1697 Supplier National Safety Apparel™ Supplier No. KIT4SC40EC5X08
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

40 cal Arc flash kit with short coat and bib overalls with gloves

  • ArcGuard™ Economy™ Short Coat and Bib Overall
  • 14 in. Class 2 Rubber Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield & hard hat
  • Safety Glasses
  • Hood bag
  • Glove bag Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 40 cal/cm²
Size 5X-Large
Suspender Adjustable
Closure Type FR Hook and Loop
Material Cotton, Nylon
Ankle Style Open
Color Tan
Seam Type Stitched
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.