Learn More
National Safety Apparel™ ArcGuard™ Compliance 40 Cal Hood Lift FRT 9 oz. Navy 2 PLY Ultrasoft
SDP
Catalog No. 17001735 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-735 Color: Navy
1 options
Catalog No. 17-001-735 Supplier National Safety Apparel™ Supplier No. H65UQUQ40LFL
Allows for quick access to fresh air and a clearer view
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 9 oz. UltraSoft Sateen, Double Layer
  • Lift front shield makes it easy to obtain fresh air while out of the hazard zone
  • Knit gussets at side provide greater range of motion and flexibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • PureView faceshield technology features a greater range of visibility
  • Hard hat included
  • Flash light with mounting clip included
  • Vents provide fresh air and increased auditory capabilities
  • Color: Navy
  • One size fits most
  • Made in the USA
Specifications
Size Medium
Material Cotton, Nylon
Color Navy
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269