Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 65 cal Arc Flash Long Coat/Legging Kit with Standard Hood/Voltage Gloves (11)
SDP

Catalog No. 170011505 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1505 Small
17-001-1510 Medium
17-001-1515 Large
17-001-1520 2X-Large
17-001-1525 3X-Large
17-001-1530 4X-Large
17-001-1538 5X-Large
17-001-1539 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1505 Supplier National Safety Apparel™ Supplier No. KIT4LC65SM11
Add to Cart
Add to Cart
Request a Quote

Constructed to be both lightweight and highly durable, this coat and leggings arc flash suit is designed for an arc flash hazard of 65 cal/cm² or less

  • FR Long Coat & Leggings
  • 14 in. Class 2 Rubber Voltage Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield and hard hat
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Material DuPont™ Nomex™, Kevlar™
Arc Flash Rating 65 cal/cm²
Suspender Adjustable
Color Brown
Ankle Style Open
Closure Type FR Hook and Loop
Seam Type Stitched
Size Small
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.