Learn More
National Safety Apparel™ ArcGuard™ Contractor 40 Cal Bib 9 oz. Khaki 2 Ply 88/12
SDP
Catalog No. 17001293 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-293 Small
17-001-294 Medium
17-001-295 Large
17-001-296 X-Large
17-001-297 2X-Large
17-001-298 3X-Large
17-001-299 4X-Large
17-001-300 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-293 Supplier National Safety Apparel™ Supplier No. C45HKHKECSM32
Add to Cart
Add to Cart
Economy bib overalls complies with OSHA 1910.269 standards.
  • 40 cal/cm²
  • 9 oz.
  • Double Layer FR Material
  • FR Fabric will not melt, drip, or ignite
  • FR hook and loop leg closure
  • Quick disconnect and easy adjustable shoulder straps for quick entry
  • Quick disconnect buckles at each shoulder
  • Inherently flame resistant for FR protection
  • Available in a convenient kit
Specifications
Closure Type Hook and Loop with Buckle
Color Tan
Material Cotton, Nylon
Seam Type Stitched
Size Small
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 40 cal/cm²
Waist Style Standard
Resistance Flame
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Disposable Reusable