Learn More
National Safety Apparel™ ArcGuard™ Contractor 40 Cal Coat 9 oz.
SDP
Catalog No. 17001277 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-277 Small
17-001-278 Medium
17-001-279 Large
17-001-280 X-Large
17-001-281 2X-Large
17-001-282 3X-Large
17-001-283 4X-Large
17-001-284 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-277 Supplier National Safety Apparel™ Supplier No. C07HKHKECSM32
Add to Cart
Add to Cart
Economy short coat that complies with OSHA 1910.269 standards.
  • 9 oz. Double Layer FR Material
  • FR Fabric will not melt, drip, or ignite
  • FR hook and loop wrist closure
  • Hook and loop front closure
  • Inherently flame resistant for FR protection
  • Available in a convient kit
Specifications
Closure Type Hook and Loop
Color Khaki
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size Small
Resistance Flame
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269