Learn More
National Safety Apparel™ ArcGuard™ Contractor 40 Cal Coat 9 oz.
SDP
Catalog No. 17001284 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-284 5X-Large
17-001-277 Small
17-001-278 Medium
17-001-279 Large
17-001-280 X-Large
17-001-281 2X-Large
17-001-282 3X-Large
17-001-283 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-284 Supplier National Safety Apparel™ Supplier No. C07HKHKEC5X32
Economy short coat that complies with OSHA 1910.269 standards.
  • 9 oz. Double Layer FR Material
  • FR Fabric will not melt, drip, or ignite
  • FR hook and loop wrist closure
  • Hook and loop front closure
  • Inherently flame resistant for FR protection
  • Available in a convient kit
Specifications
Closure Type Hook and Loop
Color Khaki
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size 5X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269