Learn More
National Safety Apparel™ ArcGuard™ Contractor 40 Cal Hood With Hat 9 oz. Khaki 2 Ply 88/12
SDP
Catalog No. 17001715 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Khaki
Catalog No.
17-001-715 Color: Khaki
1 options
Catalog No. 17-001-715 Supplier National Safety Apparel™ Supplier No. H65HKHKHATEC
Hood with slotted adapted and breathable FR fabric
  • Breathable FR fabric
  • Anti-fog faceshield
  • One size fits all
  • Slotted adapter
  • Inherently flame resistant
  • Available in a convenient kit
Specifications
Size Medium
Material Cotton, Nylon
Color Khaki
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269