Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ AGP 40 cal Arc Flash Jacket
SDP

Catalog No. 17000509 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-509 X-Small
17-000-510 Small
17-000-511 Medium
17-000-512 Large
17-000-513 X-Large
17-000-514 2X-Large
17-000-515 3X-Large
17-000-516 4X-Large
17-000-517 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-000-509 Supplier National Safety Apparel™ Supplier No. ARC40JXSR
Add to Cart
Add to Cart

AGP Arc Flash Jacket, 40 Cal, CAT 4, Back Stretch Panels, Drawstring Waist, Pleated Elbows, Knit Lined Collar

  • Single-layer 40 cal arc flash protection
  • Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits
  • Using GORE™ PYRAD™ comfort science to keep the wearer cool, dry, comfortable, and unrestricted while working
  • FR cuffs improve fit with electrical gloves
  • Covered zip front closure with large pull tab
  • Ergonomic design with added stretch back panels for ease of movement and flexibility
  • Lined collar for soft touch and moisture-wicking
  • Enhanced visibility with 3M™ Scotchlite™ FR silver segmented reflective trim

Specifications

Size X-Small
Gender Unisex
Material ePTFE, Polyamide, FR Viscose, Aramid, Carbon
Color Navy
Closure Type Zipper
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F1506
Product Type Jacket
For Use With (Application) Utilities, Electrical Contractors, Manufacturing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.