Learn More
National Safety Apparel™ ArcGuard™ Revolite Coat 40 Cal 49 in. Inseam Long
SDP
Catalog No. 17001208 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
17-001-208 X-Large
1 options
Catalog No. 17-001-208 Supplier National Safety Apparel™ Supplier No. C04NPQHLTXL49
40 cal flash coat that is 50% lighter than our standard 40 cal/cm² arc flash kits
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal/cm² arc flash kits
  • FR hook & loop front closure with drawstring waist
  • Stand up collar for extra coverage, that can be turned down
  • Nomex™ knit cuffs for a comfortable, secure fit
  • Garments embroidered with arc rating
  • FR fabric will not melt, drip, or ignite
  • Available in a convenient kit
  • Made in the USA
Specifications
Closure Type FR Hook and Loop
Color Olive Green
Disposable Reusable
Gender Unisex
Material RevoLite™
Neck Style Collared
Size X-Large
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269