Learn More
Description
2 x 3 Bottles (3.0mL each) of ARCHITECT B·R·A·H·M·S PCT L, M, and H.
- Controls. Low Control, Medium Control, and High Control contain recombinant PCT prepared in phosphate buffer. Preservatives: ProClin 950 and sodium azide.
Specifications
Specifications
| Content And Storage | Frozen at -10°C or colder. After thaw, store at 2-8°C for up to 30 days. |
| Concentration | Targeted Concentration: Control L: 0.20 ng/mL, Control M: 2.00 ng/mL, Control H: 70.00 ng/mL; Control Range: Control L: 0.14 tp 0.26 ng/mL, Control M: 1.38 to 2.62 ng/mL, Control H: 42.00 to 98.00 ng/mL |
| Type | PCT Controls |
| For Use With (Application) | Estimation of test precision and the detection of systematic analytical deviations of the ARCHITECT iSystem when used for the quantitative determination of procalcitonin (PCT) in human serum and plasma. |
| Includes | 3.0mL each of Low Control, Medium Control, and High Control |
| Intended Use | Estimation of test precision and the detection of systematic analytical deviations of the ARCHITECT iSystem when used for the quantitative determination of procalcitonin (PCT) in human serum and plasma |
| Packaging | Bottle |
| pH | 20 °C (68 °F): 6.9 |
| Quantity | 2 x 3 Bottles |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.