Learn More
Darwin Chambers Company Single Door Archival Chamber 21 cu. ft.
SDP
Catalog No. 501893918
Encompass_Preferred
The AR030 is a 23.9 cubic feet single door archival chamber. Temperature range is 0°C to 25°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. This series is capable of providing long-term controlled storage of photographs, film, rare books, papers, and paintings. Utilizing a simple precisely controlled refrigeration system and advanced desiccant-based dehumidification, temperatures are maintained to within ±0.2ºC (at the sensor) and relative humidity to within ±1% (at the sensor).
Catalog No. 50-189-3918 Supplier Darwin Chambers Company Supplier No. AR030AA
Add to Cart
Add to Cart
Request a Quote
The AR030 is a 23.9 cubic feet single door archival chamber. Temperature range is 0°C to 25°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. This series is capable of providing long-term controlled storage of photographs, film, rare books, papers, and paintings. Utilizing a simple precisely controlled refrigeration system and advanced desiccant-based dehumidification, temperatures are maintained to within ±0.2ºC (at the sensor) and relative humidity to within ±1% (at the sensor).