Learn More
Electron Microscopy Sciences PRO-LINE Archival Sleeving with Clear-Open Flap, 11x14"
SDP
Catalog No. 5031246
Encompass_Preferred
Our specially formulated 3-mil archival polypropylene has a physical density that is less than other sleeving materials, making it more pliable and gentle to your film. Strong and durable, PRO-LINE won't become brittle or abrade film. PRO-LINE sleeving is optically clear, neutral in color, and has a unique anti-Newton ring surface that reduces distracting Newton ring as well as minimizing the actual surface area that is in contact with your film.
Type: Clear-Open Flap
Size: 11 x 14"
Catalog No. 50-312-46 Supplier Electron Microscopy Sciences Supplier No. 73921CO
Our specially formulated 3-mil archival polypropylene has a physical density that is less than other sleeving materials, making it more pliable and gentle to your film. Strong and durable, PRO-LINE won't become brittle or abrade film. PRO-LINE sleeving is optically clear, neutral in color, and has a unique anti-Newton ring surface that reduces distracting Newton ring as well as minimizing the actual surface area that is in contact with your film.
Type: Clear-Open Flap
Size: 11 x 14"