Learn More
Electron Microscopy Sciences PRO-LINE Archival Sleeving Super B with Clear-Sealed Flap, 13 x 19"
SDP
Catalog No. 5031247
Encompass_Preferred
Our specially formulated 3-mil archival polypropylene has a physical density that is less than other sleeving materials, making it more pliable and gentle to your film. Strong and durable, PRO-LINE won't become brittle or abrade film. PRO-LINE sleeving is optically clear, neutral in color, and has a unique anti-Newton ring surface that reduces distracting Newton ring as well as minimizing the actual surface area that is in contact with your film.
Type: Clear-Sealed Flap
Size: 13 x 19"
Catalog No. 50-312-47 Supplier Electron Microscopy Sciences Supplier No. 73923CS
Our specially formulated 3-mil archival polypropylene has a physical density that is less than other sleeving materials, making it more pliable and gentle to your film. Strong and durable, PRO-LINE won't become brittle or abrade film. PRO-LINE sleeving is optically clear, neutral in color, and has a unique anti-Newton ring surface that reduces distracting Newton ring as well as minimizing the actual surface area that is in contact with your film.
Type: Clear-Sealed Flap
Size: 13 x 19"