Learn More
STEMCELL Technologies ArciTect™ Annealing Buffer (5X), Size: 1 mL
SDP
Catalog No. 501976234
Encompass
ArciTect™ Annealing Buffer (5X) is required to anneal ArciTect™ tracrRNA (Catalog #76016/76017/76018) and ArciTect™ crRNA (Catalog #76010/76011/76012) to form a guide RNA (gRNA) duplex. This product is a refill component for ArciTect™ tracrRNA Kit (https://www.stemcell.com/products/arcitect-tracrrna-kit.html).
Catalog No. 50-197-6234 Supplier STEMCELL Technologies Supplier No. 76020
ArciTect™ Annealing Buffer (5X) is required to anneal ArciTect™ tracrRNA (Catalog #76016/76017/76018) and ArciTect™ crRNA (Catalog #76010/76011/76012) to form a guide RNA (gRNA) duplex. This product is a refill component for ArciTect™ tracrRNA Kit (https://www.stemcell.com/products/arcitect-tracrrna-kit.html).