Learn More
STEMCELL Technologies ArciTect Cas9-eGFP Nuclease , Size: 100 ug
SDP
Catalog No. 502390377
Encompass
Enhanced green fluorescent protein (eGFP)-tagged Cas9 nuclease (3 μg/ul) for the generation of double-strand breaks in CRISPR-Cas9 genome editing
Catalog No. 50-239-0377 Supplier STEMCELL Technologies Supplier No. 76006
Enhanced green fluorescent protein (eGFP)-tagged Cas9 nuclease (3 μg/ul) for the generation of double-strand breaks in CRISPR-Cas9 genome editing