Learn More
ABclonal Technology ARCN1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A149931000UL
Encompass_Preferred
This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.
Catalog No. 50-001-05611