Learn More
Agilent Technologies ARCTIC EXPRESS DE3 COMP CELLS
Catalog No. NC9444283
Encompass
Competent cells(DE3); ArcticExpress;; Enhanced protein folding, solubility of expressed proteins; Encodes T7 RNA polymerase under control of the lacUV5 promoter; 10 x 0.1mL competent cells
Catalog No. NC9444283 Supplier Agilent Technologies Supplier No. 230192
Competent cells(DE3); ArcticExpress;; Enhanced protein folding, solubility of expressed proteins; Encodes T7 RNA polymerase under control of the lacUV5 promoter; 10 x 0.1mL competent cells