Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Arctic Express™ Dual Storage Systems
GSA_VA

Catalog No. 13262844 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 L
18.5 L
28 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-262-844 18.5 L
11-675-900 10 L
CK50922 28 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-262-844 Supplier Thermo Scientific™ Supplier No. CK50921
  • PRODUCT PROMOTION
    $4,640.00
    $2,768.00 / Each of 1

    Save $1,872.00

    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.

Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.

Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a 'can and cane' storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Manual Fill LN2 Dewar
Capacity (Metric) 18.5 L
No. of Doors 1
Model Arctic Express Dual 19
Static Holding Time 21 shipper/125 lab
Diameter (English) 14.5 in.
Diameter (English) Inside Neck 2.2 in.
Diameter (Metric) 36.8 cm
Diameter (Metric) Inside Neck 5.6 cm
Vial Capacity 175 x 2mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.