Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ARCTIC A10B Open Refrigeratiion Bath Steel
GSA_VA

Catalog No. 13875121 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
17 to 30 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-875-121 17 to 30 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-875-121 Supplier Thermo Scientific™ Supplier No. 1704108

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

For use with ARCTIC A10B Refrigeration Bath Circulator

  • Large work area enable high throughput and work flow effciency
  • Stainless steel reservoir, offered in mulitple capacities
  • Variety of reservoir openings and depth dimensions for maximum application flexibility

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Bath steel
Capacity (Metric) 17 to 30 L
Cooling Capacity 250W at 20°C
Electrical Requirements 115 V 60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.