Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Radians Arctic Radwear™ Head Shade
SDP

Catalog No. p-7221216
Change view
Click to view available options
Color:
Digital Camo
Red Paisley
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-035-282 -
19-035-284 -
19-035-285 Digital Camo
19-035-283 Red Paisley
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-035-282 Supplier Radians Supplier No. RCS305
Only null left
Add to Cart
Add to Cart

Accelerates the evaporative cooling process

  • Design allows band to move with the body for optimum blood circulation
  • For hot outdoor activities such as construction work, tough workouts or strenuous outdoor/indoor activity
  • Soak for 2 to 3 min and wear for up to 5 hrs. of cooling relief (depending on humidity)
  • Safe and reusable
  • Jobsite proven
  • Anti-microbial treated to help prevent mold build-up and unwanted smells
  • Heat stress safety management

Specifications

Product Type Head Shade
Size Universal
For Use With (Application) For hot outdoor activities such as construction work, tough workouts or any strenuous outdoor or indoor activity
Quantity 10 Pk., 50/Cs.

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.