Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Radians Arctic Radwear™ Headband
SDP

Catalog No. 19035279
Change view
Click to view available options
Color:
Blue
Blue Paisley
Hi-vis Yellow
Red Paisley
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-035-279 Hi-vis Yellow
19-035-275 Blue
19-035-278 -
19-035-277 Blue Paisley
19-035-276 Red Paisley
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-035-279 Supplier Radians Supplier No. RCS110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accelerates the evaporative cooling process

  • Safe and reusable
  • Jobsite proven
  • Anti-microbial treated to help prevent mold build-up and unwanted smells
  • Heat stress safety management
  • One size fits all
  • Soak the band in water for 2 to 3 min. and wear for up to five hours
  • Stretch-fit spandex design allows the band to move with the body
    • Allows for optimum blood circulation
  • Designed to keep cool during hot outdoor activities like construction work, tough workouts or strenuous outdoor and indoor activity

Specifications

Product Type Headband
Size Universal
Color Hi-vis Yellow
For Use With (Application) For hot outdoor activities such as construction work, tough workouts or any strenuous outdoor or indoor activity
Quantity 10 Pk., 50/Cs.

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.