Learn More
Agilent Technologies ARCTICEXPRESS DE3)RP; 10 X 0.1
Supplier:  Agilent Technologies 230194
Encompass
Enhanced protein folding and solubility of expressed proteins. Dramatically improves expression for GC-rich genomes when codon bias is a problem. Encodes T7 RNA polymerase under control of the lacUV5 promoter.
Catalog No. NC9709967