Learn More
USA Scientific Inc ArcticIce cooler, 0 deg C, white
Catalog No. NC9364620
Encompass
ArcticIce 20-place cooler for 1.5 mL or 0.5 mL tubes, maintains samples at 0C, white
Catalog No. NC9364620 Supplier USA Scientific Inc Supplier No. 23122720
ArcticIce 20-place cooler for 1.5 mL or 0.5 mL tubes, maintains samples at 0C, white