Learn More
USA Scientific Inc ArcticIce MCT, green to yellow
Catalog No. NC0348006
Encompass_Preferred
ArcticIce MCT 24-place cooler for 1.5 mL, 0.5 mL, or 2.0 mL tubes; green to yellow, 1/pack
Catalog No. NC0348006 Supplier USA Scientific Inc Supplier No. 23242726
ArcticIce MCT 24-place cooler for 1.5 mL, 0.5 mL, or 2.0 mL tubes; green to yellow, 1/pack