Learn More
Enzo Life Sciences (-)-Arctiin (25mg). CAS: 20362-31-6
SDP
Supplier:  Enzo Life Sciences ALX350318M025
Encompass_Preferred
Lignan derivative with antitumor activities. Down-regulates anti-apoptotic protein Bcl-XL. Potent inhibitor of HIV type-I integrase and DNA topoisomerase II. Shows phytoestrogenic activity and is an antagonist for platelet activating factor and Ca2+. Alternative name: (3R,4R)-(9CI)-4-[(3,4-Dimethoxyphenyl)methyl]-3-[[4-(beta-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-2(3H)-furanone. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (5mg/ml); sparingly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8841