Learn More
Arcticwhite LLC ArctiSeal 48 Well Rectangle Silicone W/ PTFE Spray Coating, Pre-scored, 48-well, Rectangle Footprint, 50 Plate Seals per Case
SDP
Catalog No. NC3862116
Encompass
ArctiSeal 48 Well Rectangle Silicone W/ PTFE Spray Coating, Pre-scored, 50 Plate Seals per Case. A sealing mat for 48-well plates with rectangular wells. It contains samples and limits evaporation and cross-contamination. Format - 48-well, rectangle footprint. Material - Silicone. Surface treatment - PTFE spray coating. Specialty - Pre-scored. Supplied by ArcticWhite as 50 plate seals per case.
Catalog No. NC3862116 Supplier Arcticwhite LLC Supplier No. AWSM2007
ArctiSeal 48 Well Rectangle Silicone W/ PTFE Spray Coating, Pre-scored, 50 Plate Seals per Case. A sealing mat for 48-well plates with rectangular wells. It contains samples and limits evaporation and cross-contamination. Format - 48-well, rectangle footprint. Material - Silicone. Surface treatment - PTFE spray coating. Specialty - Pre-scored. Supplied by ArcticWhite as 50 plate seals per case.